4-Iodo-1-methyl-1H-pyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A263384-100MG A263384-1G A263384-250MG
77000-986EA 15 USD
77000-986 77000-988 77000-990
4-Iodo-1-methyl-1H-pyrazole-3-carboxylic acid 95%
4-Iodo-1-methyl-1H-pyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₅H₅IN₂O₂
MDL Number: MFCD00465278
CAS Number: 6647-98-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR