8-Bromonaphthalen-1-amine 98%, Ambeed.Inc

Supplier: Ambeed
Warning

A263127-100MG A263127-250MG A263127-5G A263127-1G A263127-100G A263127-25G
77014-230EA 5 USD
77014-230 77014-232 77550-650 77683-802 77892-499 77892-500
8-Bromonaphthalen-1-amine 98%, Ambeed.Inc
8-Bromonaphthalen-1-amine

Precaución: For research use only.

Formula: C₁₀H₈BrN
MDL Number: MFCD01860254
CAS Number: 62456-34-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR