2-Boc-amino-3-iodo-4-chloropyridine 98%

Supplier: Ambeed
Warning

A263075-100MG A263075-1G A263075-250MG
77031-794EA 31 USD
77031-794 77031-796 77031-798
2-Boc-amino-3-iodo-4-chloropyridine 98%
2-Boc-amino-3-iodo-4-chloropyridine

Precaución: for Research Use Only

Formula: C₁₀H₁₂ClIN₂O₂
MDL Number: MFCD10687079
CAS Number: 868733-96-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR