2-Bromo-4-ethylphenol 95%

Supplier: Ambeed
Warning

A262966-1G A262966-250MG A262966-5G
77000-972EA 22 USD
77000-972 77000-974 77000-976
2-Bromo-4-ethylphenol 95%
2-Bromo-4-ethylphenol

Precaución: for Research Use Only

Formula: C₈H₉BrO
MDL Number: MFCD01593653
CAS Number: 64080-15-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR