3-Bromo-4-(hydroxymethyl)benzonitrile 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-Bromo-4-hydroxymethylbenzonitrile
A262925-100MG
A262925-1G
A262925-250MG
A262925-5G
76988-214EA
18.04
USD
76988-214
76988-216
76988-218
76988-220
3-Bromo-4-(hydroxymethyl)benzonitrile 98%
3-Bromo-4-(hydroxymethyl)benzonitrile
Precaución: for Research Use Only
Formula:
C₈H₆BrNO MW: 212.043 g/mol |
MDL Number:
MFCD09907613 CAS Number: 90110-98-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...