6-(tert-Butoxycarbonyl)-6-azaspiro[2.5]octane-1-carboxylic acid 95%

Supplier: Ambeed
Warning

Synonyms: 6-Azaspiro[2.5]octane-1-carboxylic acid, N-BOC protected

A262883-100MG A262883-1G A262883-250MG A262883-5G
77014-220EA 20.16 USD
77014-220 77014-222 77014-224 77683-801
6-(tert-Butoxycarbonyl)-6-azaspiro[2.5]octane-1-carboxylic acid 95%
6-(tert-Butoxycarbonyl)-6-azaspiro[2.5]octane-1-carboxylic acid

Precaución: For research use only.

Formula: C₁₃H₂₁NO₄
MW: 255.32 g/mol
MDL Number: MFCD11007805
CAS Number: 871727-05-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR