4,5-Dicyanoimidazole 97%

Supplier: Ambeed
Danger

Synonyms: 4,5-Dicyano-1H-imidazole, Imidazole-4,5-dicarbonitrile

A262868-25G A262868-5G A262868-10G A262868-100G A262868-1000G A262868-500G
76988-206EA 16.98 USD
76988-206 76988-208 77568-514 77571-624 77612-518 77612-520
4,5-Dicyanoimidazole 97%
4,5-Dicyanoimidazole

Precaución: For research use only.

Formula: C₅H₂N₄
MW: 118.1 g/mol
MDL Number: MFCD00005194
CAS Number: 1122-28-7
UN: 3077
ADR: 9,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR