3,5-Dimethyl-1H-pyrazole-4-carbaldehyde 95%

Supplier: Ambeed
Warning

A262858-100MG A262858-250MG A262858-1G
77031-790EA 46.68 USD
77031-790 77031-792 77571-622
3,5-Dimethyl-1H-pyrazole-4-carbaldehyde 95%
3,5-Dimethyl-1H-pyrazole-4-carbaldehyde

Precaución: for Research Use Only

Formula: C₆H₈N₂O
MDL Number: MFCD03194451
CAS Number: 201008-71-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR