3-(5-Methyl-2-furyl)propionaldehyde 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3-(5-Methyl-2-furyl)propanal
A262522-100G
A262522-1G
A262522-25G
A262522-5G
77000-964EA
610.02
USD
77000-964
77000-966
77000-968
77000-970
3-(5-Methyl-2-furyl)propionaldehyde 98%
3-(5-Methyl-2-furyl)propionaldehyde
Precaución: for Research Use Only
Formula:
C₈H₁₀O₂ MW: 138.17 g/mol |
MDL Number:
MFCD01910105 CAS Number: 34756-16-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...