1-Amino-3-methoxypropan-2-ol 95%

Supplier: Ambeed
Warning

A262464-100MG A262464-1G A262464-250MG
77031-788EA 63 USD
77031-788 77601-122 77881-713
1-Amino-3-methoxypropan-2-ol 95%
1-Amino-3-methoxypropan-2-ol

Precaución: for Research Use Only

Formula: C₄H₁₁NO₂
MDL Number: MFCD00462795
CAS Number: 93372-65-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR