Ethyl 2-amino-4-methyl-5-(4-nitrophenyl)thiophene-3-carboxylate 98%

Supplier: Ambeed
Warning

A262176-1G A262176-5G A262176-100G
77014-208EA 7 USD
77014-208 77014-210 77890-462
Ethyl 2-amino-4-methyl-5-(4-nitrophenyl)thiophene-3-carboxylate 98%
Ethyl 2-amino-4-methyl-5-(4-nitrophenyl)thiophene-3-carboxylate

Precaución: for Research Use Only

Formula: C₁₄H₁₄N₂O₄S
MDL Number: MFCD04115279
CAS Number: 174072-89-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR