2-Hydroxy-4-methoxy-5-sulfobenzophenone 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
Sulisobenzone, Benzophenone-4, 5-Benzoyl-4-hydroxy-2-methoxybenzenesulphonic acid, 2-Hydroxy-4-methoxybenzophenone-5-sulphonic Acid
A262042-10G
A262042-25G
A262042-500G
A262042-1000G
A262042-100G
77031-784EA
6.37
USD
77031-784
77031-786
77601-120
77672-338
77672-340
2-Hydroxy-4-methoxy-5-sulfobenzophenone 98%
2-Hydroxy-4-methoxy-5-sulfobenzophenone
Precaución: for Research Use Only
Formula:
C₁₄H₁₂O₆S MW: 308.30 g/mol |
MDL Number:
MFCD00024962 CAS Number: 4065-45-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...