6-Chloro-4-methyl-2,3-dihydro-1H-inden-1-one 98%
Supplier: Ambeed
|
Warning
|
A261676-100MG
A261676-1G
A261676-250MG
A261676-5G
77014-194EA
22
USD
77014-194
77014-196
77014-198
77014-200
6-Chloro-4-methyl-2,3-dihydro-1H-inden-1-one 98%
6-Chloro-4-methyl-2,3-dihydro-1H-inden-1-one
Precaución: for Research Use Only
Formula:
C₁₀H₉ClO MW: 180.63 g/mol |
MDL Number:
MFCD09908153 CAS Number: 938-35-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...