1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A261620-100MG
A261620-1G
A261620-250MG
77000-958EA
74
USD
77000-958
77000-960
77000-962
1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate hydrochloride 95%
1-Methylpyrrolidin-3-yl 2-cyclopentyl-2-hydroxy-2-phenylacetate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₈H₂₆ClNO₃ MW: 339.85 g/mol |
MDL Number:
MFCD01715893 CAS Number: 13118-10-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...