3-Hydroxy-N-(2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-naphthamide 97%

Supplier: Ambeed
Warning

A261388-1G A261388-250MG
77000-954EA 8.49 USD
77000-954 77000-956
3-Hydroxy-N-(2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-naphthamide 97%
3-Hydroxy-N-(2-oxo-2,3-dihydro-1H-benzo[d]imidazol-5-yl)-2-naphthamide

Precaución: for Research Use Only

Formula: C₁₈H₁₃N₃O₃
MW: 319.31 g/mol
MDL Number: MFCD00388332
CAS Number: 26848-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR