Methyl-3-(ethylamino)-4-nitrobenzenecarboxylate 98%

Supplier: Ambeed
Warning

A261234-1G A261234-250MG A261234-5G
77000-934EA 49 USD
77000-934 77000-936 77000-938
Methyl-3-(ethylamino)-4-nitrobenzenecarboxylate 98%
Methyl-3-(ethylamino)-4-nitrobenzenecarboxylate

Precaución: for Research Use Only

Formula: C₁₀H₁₂N₂O₄
MDL Number: MFCD09607946
CAS Number: 952183-54-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR