1-(6-Bromonaphthalen-2-yl)ethanone 97%

Supplier: Ambeed
Warning

A261151-100MG A261151-1G A261151-250MG A261151-25G A261151-5G
77014-184EA 11.67 USD
77014-184 77014-186 77014-188 77014-190 77014-192
1-(6-Bromonaphthalen-2-yl)ethanone 97%
1-(6-Bromonaphthalen-2-yl)ethanone

Precaución: for Research Use Only

Formula: C₁₂H₉BrO
MW: 249.10 g/mol
MDL Number: MFCD00126480
CAS Number: 1590-25-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR