Fmoc-3-nitro-L-tyrosine 98%

Supplier: Ambeed
Warning

A261005-1G A261005-25G A261005-5G A261005-100G
77000-930EA 14 USD
77000-930 77601-100 77601-102 77881-705
Fmoc-3-nitro-L-tyrosine 98%
Fmoc-3-nitro-L-tyrosine

Precaución: for Research Use Only

Formula: C₂₄H₂₀N₂O₇
MDL Number: MFCD00171383
CAS Number: 136590-09-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR