Ethyl 4-Methyl-2-(methylthio)-5-pyrimidinecarboxylate 97%

Supplier: Ambeed
Warning

A260949-100MG A260949-10G A260949-1G A260949-250MG A260949-25G A260949-5G
77031-766EA 22.28 USD
77031-766 77031-768 77031-770 77031-772 77031-774 77031-776
Ethyl 4-Methyl-2-(methylthio)-5-pyrimidinecarboxylate 97%
Ethyl 4-Methyl-2-(methylthio)-5-pyrimidinecarboxylate

Precaución: for Research Use Only

Formula: C₉H₁₂N₂O₂S
MW: 212.27 g/mol
MDL Number: MFCD08457402
CAS Number: 7234-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR