Sumatriptan succinate 99+%

Supplier: Ambeed
Danger

Synonyms: 1-(3-(2-(Dimethylamino)ethyl)-1H-indol-5-yl)-N-methylmethanesulphonamide succinate, 3-[2-(Dimethylamino)ethyl]-1H-indol-5-yl-N-methylmethanesulphonamide Succinate

A260899-10MG A260899-1MG
77031-758EA 6.37 USD
77031-758 77031-760 77031-762 77031-764
Sumatriptan succinate 99+%
Sumatriptan succinate

Precaución: for Research Use Only

Formula: C₁₄H₂₁N₃O₂S
MW: 413.49 g/mol
MDL Number: MFCD00902856
CAS Number: 103628-48-4
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR