2-Hydroxy-[1,1'-biphenyl]-3-carbaldehyde 95%

Supplier: Ambeed
Warning

A260853-1G A260853-5G A260853-250MG
76988-184EA 147.47 USD
76988-184 76988-186 77683-796
2-Hydroxy-[1,1'-biphenyl]-3-carbaldehyde 95%
2-Hydroxy-[1,1'-biphenyl]-3-carbaldehyde

Precaución: For research use only.

Formula: C₁₃H₁₀O₂
MW: 198.22 g/mol
MDL Number: MFCD13192635
CAS Number: 14562-10-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR