3,3-Dimethylcyclopentanone 97%

Supplier: Ambeed
Danger

A260617-1G A260617-250MG A260617-5G
76988-178EA 38 USD
76988-178 76988-180 76988-182
3,3-Dimethylcyclopentanone 97%
3,3-Dimethylcyclopentanone

Precaución: for Research Use Only

Formula: C₇H₁₂O
MDL Number: MFCD18917067
CAS Number: 20500-49-6
UN: 1224
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR