Benzofurazan-5-boronic acid pinacol ester 98+%

Supplier: Ambeed
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzofurazan, 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzo[c][1,2,5]oxadiazole

A260597-1G A260597-250MG A260597-5G
77031-752EA 63 USD
77031-752 77031-754 77031-756
Benzofurazan-5-boronic acid pinacol ester 98+%
Benzofurazan-5-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₅BN₂O₃
MDL Number: MFCD09842717
CAS Number: 1073355-14-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR