2-Methoxy-3-nitropyridine 97%

Supplier: Ambeed
Warning

A260561-100G A260561-25G A260561-5G
77000-924EA 266.29 USD
77000-924 77000-926 77000-928
2-Methoxy-3-nitropyridine 97%
2-Methoxy-3-nitropyridine

Precaución: for Research Use Only

Formula: C₆H₆N₂O₃
MDL Number: MFCD00023459
CAS Number: 20265-35-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR