2-Oxa-8-azaspiro[4.5]decan-1-one hydrochloride 97%

Supplier: Ambeed
Warning

A260327-100MG A260327-250MG
77031-746EA 37.14 USD
77031-746 77031-748
2-Oxa-8-azaspiro[4.5]decan-1-one hydrochloride 97%
2-Oxa-8-azaspiro[4.5]decan-1-one hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₄ClNO₂
MW: 191.66 g/mol
MDL Number: MFCD11035931
CAS Number: 1314965-08-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR