4-Bromo-3,5-difluorobenzonitrile 97%

Supplier: Ambeed
Warning

A260212-1G A260212-5G A260212-25G A260212-250MG
77014-176EA 67.9 USD
77014-176 77014-178 77593-624 77683-793
4-Bromo-3,5-difluorobenzonitrile 97%
4-Bromo-3,5-difluorobenzonitrile

Precaución: For research use only.

Formula: C₇H₂BrF₂N
MW: 217.9983 g/mol
MDL Number: MFCD11845964
CAS Number: 123688-59-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR