2-(Aminomethyl)-1,3-oxazole hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (1,3-Oxazol-2-yl)methylamine hydrochloride, Oxazol-2-ylmethanamine hydrochloride

A260074-1G A260074-250MG A260074-100MG
77014-170EA 226 USD
77014-170 77014-172 77612-488
2-(Aminomethyl)-1,3-oxazole hydrochloride 98%
2-(Aminomethyl)-1,3-oxazole hydrochloride

Precaución: for Research Use Only

Formula: C₄H₆N₂O·ClH
MDL Number: MFCD06738929
CAS Number: 1041053-44-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR