Candesartan cilexetil 98%

Supplier: Ambeed
Danger

Synonyms: 1-(((Cyclohexyloxy)carbonyl)oxy)ethyl 1-((2'-(2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-2-ethoxy-1H-benzo[d]imidazole-7-carboxylate

A259269-10MG A259269-1G A259269-1MG A259269-25G A259269-50MG A259269-5G A259269-10G
77000-902EA 6.37 USD
77000-902 77000-904 77000-906 77000-908 77000-910 77593-620 77672-314
Candesartan cilexetil 98%
Candesartan cilexetil

Precaución: for Research Use Only

Formula: C₃₃H₃₄N₆O₆
MW: 610.67 g/mol
MDL Number: MFCD00871371
CAS Number: 145040-37-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR