5-tert-Butyl-1,3,4-oxadiazol-2-amine 97%

Supplier: Ambeed
Warning

Synonyms: 5-(tert-Butyl)-1,3,4-oxadiazol-2-amine

A259152-100MG A259152-1G A259152-250MG
77031-736EA 26.53 USD
77031-736 77031-738 77031-740
5-tert-Butyl-1,3,4-oxadiazol-2-amine 97%
5-tert-Butyl-1,3,4-oxadiazol-2-amine

Precaución: for Research Use Only

Formula: C₆H₁₁N₃O
MW: 141.17 g/mol
MDL Number: MFCD09261284
CAS Number: 69741-92-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR