(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-carbamoyl-2,6-dimethylphenyl)propanoic acid 98%

Supplier: Ambeed
Warning

A258975-1G A258975-250MG A258975-5G
77014-156EA 10 USD
77014-156 77014-158 77014-160
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-carbamoyl-2,6-dimethylphenyl)propanoic acid 98%
(S)-2-((tert-Butoxycarbonyl)amino)-3-(4-carbamoyl-2,6-dimethylphenyl)propanoic acid

Precaución: for Research Use Only

Formula: C₁₇H₂₄N₂O₅
MDL Number: MFCD20528368
CAS Number: 623950-02-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR