3-Chloro-4-(2'-chlorobenzyloxy)phenylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (3-Chloro-4-((2-chlorobenzyl)oxy)phenyl)boronic acid

A258678-1G A258678-250MG A258678-5G
77014-150EA 87 USD
77014-150 77014-152 77014-154
3-Chloro-4-(2'-chlorobenzyloxy)phenylboronic acid 95%
3-Chloro-4-(2'-chlorobenzyloxy)phenylboronic acid

Precaución: for Research Use Only

Formula: C₁₃H₁₁BCl₂O₃
MW: 296.9400 g/mol
MDL Number: MFCD06798081
CAS Number: 870777-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR