Dimethyl fumarate 99%

Supplier: Ambeed
Warning

Synonyms: Fumaric acid dimethyl ester

A258585-1000G A258585-100G A258585-10G A258585-25G A258585-500G
77014-140EA 45 USD
77014-140 77014-142 77014-144 77014-146 77014-148
Dimethyl fumarate 99%
Dimethyl fumarate

Precaución: for Research Use Only

Formula: C₆H₈O₄
MW: 144.13 g/mol
Boiling Pt: 192
Melting Pt: 102
MDL Number: MFCD00064438
CAS Number: 624-49-7
Merck Index: 13,04308

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR