3-Bromo-6-fluoro-2-methoxybenzeneboronic acid 97%

Supplier: Ambeed
Warning

Synonyms: (3-Bromo-6-fluoro-2-methoxyphenyl)boronic acid, 3-Bromo-6-fluoro-2-methoxyphenylboronic acid

A258526-100MG A258526-1G A258526-250MG
77000-894EA 5.31 USD
77000-894 77000-896 77000-898
3-Bromo-6-fluoro-2-methoxybenzeneboronic acid 97%
3-Bromo-6-fluoro-2-methoxybenzeneboronic acid

Precaución: for Research Use Only

Formula: C₇H₇BBrFO₃
MW: 248.842 g/mol
MDL Number: MFCD09475911
CAS Number: 957120-30-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR