Methyl 3-bromoimidazo[1,2-a]pyridine-8-carboxylate 97%

Supplier: Ambeed
Warning

A258279-100MG A258279-1G A258279-250MG
76988-122EA 12 USD
76988-122 76988-124 76988-126
Methyl 3-bromoimidazo[1,2-a]pyridine-8-carboxylate 97%
Methyl 3-bromoimidazo[1,2-a]pyridine-8-carboxylate

Precaución: for Research Use Only

Formula: C₉H₇BrN₂O₂
MW: 255.06 g/mol
MDL Number: MFCD13195318
CAS Number: 1234616-47-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR