N3-(3-Bromobenzyl)pyridine-2,3-diamine 95%

Supplier: Ambeed
Warning

A258007-1G A258007-250MG
77031-718EA 39.26 USD
77031-718 77031-720
N3-(3-Bromobenzyl)pyridine-2,3-diamine 95%
N3-(3-Bromobenzyl)pyridine-2,3-diamine

Precaución: for Research Use Only

Formula: C₁₂H₁₂BrN₃
MW: 278.15 g/mol
MDL Number: MFCD09701365
CAS Number: 934537-52-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR