2-(1H-Imidazol-2-yl)ethanamine dihydrochloride 98%

Supplier: Ambeed
Warning

A257997-100MG A257997-1G A257997-250MG
77014-118EA 77.45 USD
77014-118 77014-120 77014-122
2-(1H-Imidazol-2-yl)ethanamine dihydrochloride 98%
2-(1H-Imidazol-2-yl)ethanamine dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₁Cl₂N₃
MW: 184.06 g/mol
MDL Number: MFCD08668396
CAS Number: 17286-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR