4-Chloro-2,5-difluoropyridine 95%

Supplier: Ambeed
Warning

A257956-100MG A257956-1G A257956-250MG A257956-5G
77000-886EA 45.62 USD
77000-886 77000-888 77000-890 77000-892
4-Chloro-2,5-difluoropyridine 95%
4-Chloro-2,5-difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₂ClF₂N
MDL Number: MFCD09031178
CAS Number: 851386-40-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR