Ranitidine hydrochloride 97%

Supplier: Ambeed
Danger

Synonyms: N-[2-[5-[(Dimethylamino)methyl]furfurylthio]ethyl]-N'-methyl-2-nitro-1,1-ethenediamine Hydrochloride, N-(2-(((5-((Dimethylamino)methyl)furan-2-yl)methyl)thio)ethyl)-N'-methyl-2-nitroethene-1,1-diamine hydrochloride

A257429-100G A257429-100MG A257429-1G A257429-25G A257429-500G A257429-5G A257429-1MG A257429-10G
76988-100EA 173 USD
76988-100 76988-102 76988-104 76988-106 76988-108 76988-110 77612-470 77885-674
Ranitidine hydrochloride 97%
Ranitidine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₂₂N₄O₃S
MDL Number: MFCD00069339
CAS Number: 66357-59-3
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR