6,6'-Dichloro-2,2'-bipyridine 97%, Ambeed.Inc

Supplier: Ambeed
Warning

A257383-100MG A257383-250MG A257383-5G A257383-1G
76988-096EA 16 USD
76988-096 76988-098 77683-784 77683-783
6,6'-Dichloro-2,2'-bipyridine 97%, Ambeed.Inc
6,6'-Dichloro-2,2'-bipyridine

Precaución: For research use only.

Formula: C₁₀H₆Cl₂N₂
MW: 225.07 g/mol
MDL Number: MFCD11042483
CAS Number: 53344-72-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR