Bis(2-[di(1-adamantyl)phosphino]ethyl)amine 98%

Supplier: Ambeed
Danger

Synonyms: Bis(2-(Di(adamantan-1-yl)phosphino)ethyl)amine

A257374-100MG A257374-1G A257374-250MG
76988-090EA 165 USD
76988-090 76988-092 76988-094
Bis(2-[di(1-adamantyl)phosphino]ethyl)amine 98%
Bis(2-[di(1-adamantyl)phosphino]ethyl)amine

Precaución: for Research Use Only

Formula: C₄₄H₆₉NP₂
MDL Number: MFCD17018779
CAS Number: 1086138-36-4
UN: 3259
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR