6-(2-(tert-Butoxycarbonyl)hydrazinyl)nicotinic acid 95%
Supplier: Ambeed
|
Warning
|
A257341-100MG
A257341-10G
A257341-1G
A257341-250MG
A257341-5G
77000-856EA
19.1
USD
77000-856
77000-858
77000-860
77000-862
77000-864
6-(2-(tert-Butoxycarbonyl)hydrazinyl)nicotinic acid 95%
6-(2-(tert-Butoxycarbonyl)hydrazinyl)nicotinic acid
Precaución: for Research Use Only
Formula:
C₁₁H₁₅N₃O₄ MW: 253.25 g/mol |
MDL Number:
MFCD03788696 CAS Number: 133081-25-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...