6-Fluoronicotinic acid tert-butyl ester 95%

Supplier: Ambeed
Warning

Synonyms: tert-Butyl 6-fluoronicotinate

A257254-100MG A257254-1G A257254-250MG A257254-5G
76988-078EA 22.28 USD
76988-078 76988-080 76988-082 76988-084
6-Fluoronicotinic acid tert-butyl ester 95%
6-Fluoronicotinic acid tert-butyl ester

Precaución: for Research Use Only

Formula: C₁₀H₁₂FNO₂
MW: 197.206 g/mol
MDL Number: MFCD26743533
CAS Number: 676560-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR