4-Nitro-α,α,α-trifluoro-m-toluic acid 98%

Supplier: Ambeed
Warning

A257152-10G A257152-1G A257152-250MG A257152-5G
76988-070EA 109 USD
76988-070 76988-072 76988-074 76988-076
4-Nitro-α,α,α-trifluoro-m-toluic acid 98%
4-Nitro-α,α,α-trifluoro-m-toluic acid

Precaución: for Research Use Only

Formula: C₈H₄F₃NO₄
MDL Number: MFCD06200716
CAS Number: 320-38-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR