Ethyl 2-chloro-4-fluorobenzoate 95+%

Supplier: Ambeed
Warning

A256572-100G A256572-25G A256572-5G
76988-054EA 59 USD
76988-054 76988-056 76988-058
Ethyl 2-chloro-4-fluorobenzoate 95+%
Ethyl 2-chloro-4-fluorobenzoate

Precaución: for Research Use Only

Formula: C₉H₈ClFO₂
MDL Number: MFCD06204388
CAS Number: 167758-87-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR