1,3-Benzothiazole-6-sulfonyl chloride 96%

Supplier: Ambeed
Danger

Synonyms: Benzo[d]thiazole-6-sulphonyl chloride, 6-(Chlorosulphonyl)-1,3-benzothiazole

A256184-100MG A256184-10G A256184-250MG A256184-5G A256184-1G
77000-820EA 22.28 USD
77000-820 77000-822 77000-824 77000-826 77593-602
1,3-Benzothiazole-6-sulfonyl chloride 96%
1,3-Benzothiazole-6-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₇H₄ClNO₂S₂
MW: 233.695 g/mol
MDL Number: MFCD03086100
CAS Number: 181124-40-3
UN: 1759
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR