N-Boc-3-cyano-D-phenylalanine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
Boc-D-Phe(3-CN)-OH
A256125-1G
A256125-250MG
A256125-5G
77000-812EA
35
USD
77000-812
77000-814
77000-816
N-Boc-3-cyano-D-phenylalanine 95%
N-Boc-3-cyano-D-phenylalanine
Precaución: for Research Use Only
Formula:
C₁₅H₁₈N₂O₄ MW: 290.319 g/mol |
MDL Number:
MFCD00797561 CAS Number: 205445-56-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...