tert-Butyl 3-hydroxybenzoate 96%

Supplier: Ambeed
Warning

A256012-250MG A256012-1G
76988-038EA 55.17 USD
76988-038 77683-778
tert-Butyl 3-hydroxybenzoate 96%
tert-Butyl 3-hydroxybenzoate

Precaución: For research use only.

Formula: C₁₁H₁₄O₃
MW: 194.227 g/mol
MDL Number: MFCD09953071
CAS Number: 57704-54-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR