3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-1H-pyrazolo[3,4-b]pyridin-3-amine

A256004-100MG A256004-1G A256004-250MG A256004-25G A256004-5G
76988-028EA 6.37 USD
76988-028 76988-030 76988-032 76988-034 76988-036
3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine 97%
3-Amino-5-fluoro-1H-pyrazolo[3,4-b]pyridine

Precaución: for Research Use Only

Formula: C₆H₅FN₄
MW: 152.13 g/mol
MDL Number: MFCD15526909
CAS Number: 1034667-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR