Methyl-2-chloro-5-methylnicotinate 97%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-chloro-5-methylpyridine-3-carboxylate, Methyl 2-chloro-5-methylnicotinate

A255927-1G A255927-250MG
76988-026EA 60 USD
76988-026 77890-447
Methyl-2-chloro-5-methylnicotinate 97%
Methyl-2-chloro-5-methylnicotinate

Precaución: for Research Use Only

Formula: C₈H₈ClNO₂
MDL Number: MFCD07375372
CAS Number: 65169-43-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR