1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one 95+%

Supplier: Ambeed
Warning

Synonyms: 1-Cyclopropyl-3-(dimethylamino)prop-2-en-1-one

A255857-1G A255857-250MG A255857-5G
77000-806EA 122.01 USD
77000-806 77000-808 77000-810
1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one 95+%
1-Cyclopropyl-3-(dimethylamino)-2-propen-1-one

Precaución: for Research Use Only

Formula: C₈H₁₃NO
MW: 139.2 g/mol
MDL Number: MFCD11840950
CAS Number: 21666-68-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR